Reagents Methyl Red Indicator, 0.02% (w/v) Aqueous, Amber Glass Bottle, 125 mL

Representative image only.

Reagents Methyl Red Indicator, 0.02% (w/v) Aqueous, Amber Glass Bottle, 125 mL

Reagents | Mfr: C7118600-125C| Item: 6228770

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Aqueous
  • Orange to dark red liquid
  • Solubility information: miscible
  • pH range: 4.2 (pink) to 6.2 (yellow)
  • 125 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.02% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)125
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionMethyl Red Indicator, 0.02% (w/v) Aqueous, Amber Glass Bottle, 125 mL
Documents & Certificates
Reviews
Q&A