Reagents Methyl Ethyl Ketone, ACS Reagent, Amber Glass Bottle, 1 L

Representative image only.

Reagents Methyl Ethyl Ketone, ACS Reagent, Amber Glass Bottle, 1 L

Reagents | Mfr: C1250500-1C| Item: 6228220

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless liquid
  • Density: 0.81
  • Flash point: -9 °C (15.8 °F)
  • Refractive index: 1.37 to 1.38nD
  • Solubility information: 27.1 g/L at 20 °C (68 °F)
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCH3COCH2CH3
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Butanone, Ethyl methyl ketone, MEK
  • Volume (mL)1000
  • Formula Weight72.11
  • Vapor Pressure (torr)75.756
  • PackagingAmber glass bottle
  • DOT InformationMETHYL ETHYL KETONE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number78-93-3
  • UN NumberUN1193
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)79.6
  • Boiling Point F (° F)175.28
  • Melting Point C (° C)-86.4
  • Melting Point F (° F)-123.52
  • DescriptionMethyl Ethyl Ketone, ACS Reagent, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A