Reagents Methanol, Synthetic, Drum, 200 L

Representative image only.

Reagents Methanol, Synthetic, Drum, 200 L

Reagents | Mfr: C2121000-200E| Item: 6228378

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.79
  • Boiling point: approximately 64.5 °C (148.1 °F)
  • Flash point: 11 °C (51.8 °F)
  • Refractive index: 1.328 to 1.33nD
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaCH3OH
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsMethyl alcohol
  • FormLiquid
  • Volume (mL)200000
  • Formula Weight32.04
  • Vapor Pressure (torr)96.0079
  • PackagingDrum
  • DOT InformationMETHANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-56-1
  • UN NumberUN1230
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionMethanol, Synthetic, Drum, 200 L
Documents & Certificates
Reviews
Q&A