Reagents Magnesium Hydroxide, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Magnesium Hydroxide, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2238700-500A5| Item: 6228522

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.36
  • Solubility information: 0.009 g/L at 18 °C (64.4 °F)
  • 500 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaMg(OH)2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)500
  • Formula Weight58.32
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1309-42-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)350
  • Melting Point F (° F)662
  • DescriptionMagnesium Hydroxide, Purified, Wide-Mouth Container, 500 g
Documents & Certificates
Reviews
Q&A