Reagents Lead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Lead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1229000-100C2| Item: 6228207

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum 99%
  • Colorless/white solid
  • Density: 4.53
  • Melting point: 470 °C (878 °F), decomposes
  • Solubility information: 565 g/L at 20 °C (68 °F)
  • 100 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaPb(NO3)2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)100
  • Formula Weight331.21
  • PackagingWide-mouth amber glass bottle
  • DOT InformationLEAD NITRATE, 5.1 (6.1), P.G. II
  • Storage SymbolAmbient
  • CAS Number10099-74-8
  • UN NumberUN1469
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionLead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
Documents & Certificates
Reviews
Q&A