Reagents Lead Acetate Trihydrate, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Lead Acetate Trihydrate, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1226500-12KG| Item: 6228206

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum 99%
  • Colorless/white solid
  • Density: 2.55
  • Boiling point: 100 °C (212 °F), decomposes
  • Solubility information: very soluble
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(CH3COO)2Pb·3H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSugar of lead
  • FormSolid
  • Weight (g)12000
  • PackagingPoly pail
  • DOT InformationLEAD ACETATE, 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1616
  • CAS Number6080-56-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)75
  • Melting Point F (° F)167
  • DescriptionLead Acetate Trihydrate, ACS Reagent, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A