Reagents Lanthanum Oxide, Reagent, Wide-Mouth Container, 1 kg

Representative image only.

Reagents Lanthanum Oxide, Reagent, Wide-Mouth Container, 1 kg

Reagents | Mfr: C2224350-1KG| Item: 6228518

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 6.51
  • Boiling point: approximately 4200 °C (759217 °F)
  • Solubility information: insoluble
  • 1 kg wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaLa2O3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)1000
  • Formula Weight325.81
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1312-81-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)2315
  • Melting Point F (° F)4199
  • DescriptionLanthanum Oxide, Reagent, Wide-Mouth Container, 1 kg
Documents & Certificates
Reviews
Q&A