Reagents Lactic Acid, 85%, Purified, Natural Poly Container, 100 mL

Representative image only.

Reagents Lactic Acid, 85%, Purified, Natural Poly Container, 100 mL

Reagents | Mfr: C2108000-100A| Item: 6228349

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 1.21
  • Solubility information: miscible
  • 100 mL natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.85
  • Purity GradePurified
  • Linear FormulaCH3CHOHCOOH
  • ClassAcid
  • FormLiquid
  • Viscosity (cSt)36.9
  • Volume (mL)100
  • Formula Weight90.08
  • Vapor Pressure (torr)0.0813
  • PackagingPoly containerottle
  • DOT InformationCORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. (LACTIC ACID), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number50-21-5
  • UN NumberUN3265
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)122
  • Boiling Point F (° F)251.6
  • Melting Point C (° C)17
  • Melting Point F (° F)62.6
  • DescriptionLactic Acid, 85%, Purified, Natural Poly Container, 100 mL
Documents & Certificates
Reviews
Q&A