Reagents Isopropanol, ACS Reagent, Natural Poly Bottle, 4 L

Representative image only.

Reagents Isopropanol, ACS Reagent, Natural Poly Bottle, 4 L

Reagents | Mfr: C1121500-4A| Item: 6228135

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 0.78
  • Flash point: 12 °C (53.6 °F)
  • Refractive index: 1.376 to 1.378nD
  • 4 L natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCH3CHOHCH3
  • ClassSolvents
  • ChemistrySolvents
  • Synonyms2-Propanol, Dimethyl carbinol, IPA, Isopropyl alcohol, Isopropanol, Rubbing alcohol
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight60.1
  • Vapor Pressure (torr)31.502
  • PackagingPoly bottle
  • DOT InformationISOPROPANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-63-0
  • UN NumberUN1219
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)82.5
  • Boiling Point F (° F)180.5
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • DescriptionIsopropanol, ACS Reagent, Natural Poly Bottle, 4 L
Documents & Certificates
Reviews
Q&A