Reagents Iodine, ACS Reagent, Natural Poly Container, 2 kg

Representative image only.

Reagents Iodine, ACS Reagent, Natural Poly Container, 2 kg

Reagents | Mfr: C1217500-2KG| Item: 6228204

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.8%
  • Violet-black crystal
  • Density: 4.93
  • Solubility information: 3 g/L at 20 °C (68 °F)
  • Natural poly container
  • 2 kg

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaI2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)2000
  • PackagingPoly container
  • DOT InformationIODINE, 8 (6.1), P.G. III
  • Storage SymbolAmbient
  • CAS Number7553-56-2
  • UN NumberUN3495
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)184
  • Boiling Point F (° F)363.2
  • Melting Point C (° C)114
  • Melting Point F (° F)237.2
  • DescriptionIodine, ACS Reagent, Natural Poly Container, 2 kg
Documents & Certificates
Reviews
Q&A