Reagents Hydrochloric Acid, 20° Baumé, Technical, Drum, 200 L

Representative image only.

Reagents Hydrochloric Acid, 20° Baumé, Technical, Drum, 200 L

Reagents | Mfr: C5101000-200E| Item: 6228635

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly greenish-yellow liquid
  • Density: 1.15
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately -28 °C (-18.4 °F)
  • Solubility information: miscible
  • pH range: <1
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaHCl
  • ClassAcid
  • FormLiquid
  • SynonymsMuriatic acid, Hydrogen chloride
  • Volume (mL)20000
  • Formula Weight36.46
  • PackagingDrum
  • DOT InformationHYDROCHLORIC ACID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number7647-01-0
  • UN NumberUN1789
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionHydrochloric Acid, 20° Baumé, Technical, Drum, 200 L
Documents & Certificates
Reviews
Q&A