Reagents Hexane(s), ACS Reagent, Drum, 15 gal.

Representative image only.

Reagents Hexane(s), ACS Reagent, Drum, 15 gal.

Reagents | Mfr: C1213900-15E| Item: 6228199

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 95% n-Hexane
  • Colorless liquid
  • Density: 0.66
  • Flash point: –23 °C (–9.4 °F)
  • Refractive index: 1.374 to 1.381nD
  • Solubility information: 0.0095 g/L at 20 °C (68 °F)
  • 15 gallon drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6H14
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (Gal)15
  • PackagingDrum
  • DOT InformationHEXANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number110-54-3
  • UN NumberUN1208
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)69
  • Boiling Point F (° F)156.2
  • Melting Point C (° C)-95
  • Melting Point F (° F)-139
  • DescriptionHexane(s), ACS Reagent, Drum, 15 gal.
Documents & Certificates
Reviews
Q&A