Reagents Heptanes, Technical, Amber Glass Bottle, 4 L

Representative image only.

Reagents Heptanes, Technical, Amber Glass Bottle, 4 L

Reagents | Mfr: C2213000-4C| Item: 6228508

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.68
  • Flash point: -4 °C (24.8 °F)
  • Refractive index: 1.385 to 1.389nD
  • Solubility information: 3 µg/mL at 20 °C (68 °F)
  • 4 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaC7H16
  • ClassSolvents
  • ChemistrySolvents
  • Synonymsn-Heptane, normal heptane, Dipropyl methane, Heptyl hydride
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight100.2
  • Vapor Pressure (torr)39.7533
  • PackagingAmber glass bottle
  • DOT InformationHEPTANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number142-82-5
  • UN NumberUN1206
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)98.4
  • Boiling Point F (° F)209.12
  • Melting Point C (° C)-90.6
  • Melting Point F (° F)-131.08
  • DescriptionHeptanes, Technical, Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A