Reagents Glycerol, ACS Reagent, Drum, 200 L

Representative image only.

Reagents Glycerol, ACS Reagent, Drum, 200 L

Reagents | Mfr: C1210500-200E| Item: 6228197

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 1.26
  • Boiling point: 544 °C (1011.2 °F), decomposes
  • Flash point: 199 °C (390.2 °F)
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC3H8O3
  • ClassAlcohol
  • ChemistrySolvents
  • FormLiquid
  • SynonymsGlycerin, Propanetriol
  • Volume (mL)200000
  • Formula Weight92.09
  • Vapor Pressure (torr)0.00248
  • PackagingDrum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number56-81-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)18.2
  • Melting Point F (° F)64.76
  • Refractive Index1.4716
  • DescriptionGlycerol, ACS Reagent, Drum, 200 L
Documents & Certificates
Reviews
Q&A