Reagents Formic Acid, 90%, Viscosity Grade, Drum, 200 L

Representative image only.

Reagents Formic Acid, 90%, Viscosity Grade, Drum, 200 L

Reagents | Mfr: C2105050-200E| Item: 6228348

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Low iron
  • Colorless liquid
  • Density: 1.22
  • Flash point: 50 °C (122 °F)
  • Solubility information: miscible
  • pH range: 1.2
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.9
  • Purity GradeViscosity
  • Linear FormulaHCOOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)200000
  • Formula Weight46.03
  • Vapor Pressure (torr)30.0025
  • PackagingDrum
  • DOT InformationFORMIC ACID, 8 (3), P.G. II
  • Storage SymbolAmbient
  • CAS Number64-18-6
  • UN NumberUN1779
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)101
  • Boiling Point F (° F)213.8
  • DescriptionFormic Acid, 90%, Viscosity Grade, Drum, 200 L
Documents & Certificates
Reviews
Q&A