Reagents Formic Acid, 90%, Purified, Clear Glass Bottle, 4 L

Representative image only.

Reagents Formic Acid, 90%, Purified, Clear Glass Bottle, 4 L

Reagents | Mfr: C2105000-4A| Item: 6228346

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless, liquid
  • Density: 1.22
  • Boiling point: approximately 101 °C (213.8 °F)
  • Flash point: 50 °C (122 °F)
  • Solubility information: miscible
  • pH range: 1.2
  • 4 L clear glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.9
  • Purity GradePurified
  • Linear FormulaHCOOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight46.03
  • PackagingClear glass bottle
  • DOT InformationFORMIC ACID, 8 (3), P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1779
  • CAS Number64-18-6
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionFormic Acid, 90%, Purified, Clear Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A