Reagents Florisil®, 60 to 100 Mesh, Grade A, Poly Pail, 3 kg

Representative image only.

Reagents Florisil®, 60 to 100 Mesh, Grade A, Poly Pail, 3 kg

Reagents | Mfr: C2208300-3KG| Item: 6228506

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White powder
  • Density: 2.51
  • Mesh size: 60 to 100
  • Solubility information: insoluble
  • 3 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeA
  • Linear FormulaMgO:xSiO2·H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsFlorisil R
  • FormSolid
  • Weight (g)3000
  • Formula Weight100.38
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1343-88-0
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionFlorisil®, 60 to 100 Mesh, Grade A, Poly Pail, 3 kg
Documents & Certificates
Reviews
Q&A