Reagents Ferrous Chloride, Tetrahydrate, Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Ferrous Chloride, Tetrahydrate, Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C1206500-500C2| Item: 6228193

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Green to blue-green solid
  • Density: 1.93
  • Boiling point: sublimes
  • Melting point: (anhydrous) 607 to 674 °C (1238 to 1245.2 °F)
  • Solubility information: 650 g/L at 25 °C (77 °F)
  • 500 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeReagent
  • Linear FormulaFeCl2·4H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsIron(II) chloride 4-hydrate, Iron(II) chloride tetrahydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight198.81
  • PackagingWide-mouth amber glass bottle
  • DOT InformationFERROUS CHLORIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number13478-10-9
  • UN NumberNA1759
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionFerrous Chloride, Tetrahydrate, Reagent, Wide-Mouth Amber Glass Bottle, 500 g
Documents & Certificates
Reviews
Q&A