Reagents Ferrous Ammonium Sulfate, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Ferrous Ammonium Sulfate, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1206000-12KG| Item: 6228190

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98.5%
  • Pale blue-green solid
  • Density: 1.86
  • Melting point: 100 to 110 °C (212 to 230 °F), decomposes
  • Solubility information: 269 g/L at 20 °C (68 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaFe(NH4)2(SO4)2·6H20
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)12000
  • Formula Weight392.14
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7783-85-9
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionFerrous Ammonium Sulfate, ACS Reagent, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A