Reagents Ferric Chloride Hexahydrate, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Ferric Chloride Hexahydrate, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1204000-12KG| Item: 6228185

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Brownish-orange solid
  • Density: 2.9
  • Boiling point: 280 to 285 °C (536 to 545 °F)
  • Solubility information: 912 g/L at 25 °C (77 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaFeCl3·6H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsFerric chloride, Iron (III) chloride, Iron trichloride, Iron(III) chloride hexahydrate
  • Weight (g)12000
  • Formula Weight270.2957
  • Vapor Pressure (torr)< 0.0075
  • PackagingPoly pail
  • Melting Point C (° C)37
  • Melting Point F (° F)98.6
Documents & Certificates
Reviews
Q&A