Reagents Ethylene Glycol (1,2-Ethanediol; Ethylene Alcohol), 4L

Representative image only.

Reagents Ethylene Glycol (1,2-Ethanediol; Ethylene Alcohol), 4L

Reagents | Mfr: C2201500-4F| Item: 6228503

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless viscous liquid
  • Density: 1.11
  • Flash point: 111 °C (231.8 °F)
  • Refractive index: 1.43 to 1.433nD
  • Solubility information: miscible

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaHOCH2CH2OH
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsEthane-1,2-diol, Monoethylene glycol, 1,2-Ethanediol, Ethylene alcohol
  • Volume (mL)4000
  • Formula Weight62.07
  • Vapor Pressure (torr)0.07501
  • PackagingCubitainer®
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number107-21-1
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)198
  • Boiling Point F (° F)388.4
  • Melting Point C (° C)-13
  • Melting Point F (° F)8.6
  • DescriptionEthylene Glycol, Purified, Cubitainer® Package, 4 L
Documents & Certificates
Reviews
Q&A