Reagents Ethyl Alcohol, Denatured, 95%, Natural Poly Container, 1 L

Representative image only.

Reagents Ethyl Alcohol, Denatured, 95%, Natural Poly Container, 1 L

Reagents | Mfr: C2120500-1A| Item: 6228370

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 190 proof
  • Colorless liquid
  • Density: 0.8
  • Boiling point: approximately 77 °C (170.6 °F)
  • Melting point: approximately -90 °C (-130 °F)
  • Flash point: 14 °C (57.2 °F)
  • Solubility information: miscible
  • 1 L natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.95
  • ClassSolvents
  • ChemistrySolvents
  • Proof190
  • FormLiquid
  • Volume (mL)1000
  • Vapor Pressure (torr)48.4
  • PackagingPoly container
  • DOT InformationALCOHOLS, N.O.S. (ETHANOL, METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number64-17-5
  • UN NumberUN1987
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionEthyl Alcohol, Denatured, 95%, Natural Poly Container, 1 L
Documents & Certificates
Reviews
Q&A