Reagents Ethyl Alcohol, Denatured, 95%, Drum, 200 L

Representative image only.

Reagents Ethyl Alcohol, Denatured, 95%, Drum, 200 L

Reagents | Mfr: C2120500-200E| Item: 6228371

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 190 proof
  • Colorless liquid
  • Density: 0.8
  • Boiling point: approximately 77 °C (170.6 °F)
  • Melting point: approximately -90 °C (-130 °F)
  • Flash point: 14 °C (57.2 °F)
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.95
  • ClassSolvents
  • ChemistrySolvents
  • Proof190
  • FormLiquid
  • Volume (mL)200000
  • Vapor Pressure (torr)48.4
  • PackagingDrum
  • DOT InformationALCOHOLS, N.O.S. (ETHANOL, METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1987
  • CAS Number64-17-5
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionEthyl Alcohol, Denatured, 95%, Drum, 200 L
Documents & Certificates
Reviews
Q&A