Reagents Ethyl Alcohol, Denatured, 70%, Cubitainer® Package, 20 L

Representative image only.

Reagents Ethyl Alcohol, Denatured, 70%, Cubitainer® Package, 20 L

Reagents | Mfr: C2120400-20F| Item: 6228369

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 140 proof
  • Colorless liquid
  • Density: 0.89
  • Flash point: 21 °C (69.8 °F)
  • Solubility information: miscible
  • 20 L Cubitainer package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.7
  • ClassSolvents
  • ChemistrySolvents
  • Proof140
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer®
  • DOT InformationETHANOL SOLUTION, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number64-17-5
  • UN NumberUN1170
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionEthyl Alcohol, Denatured, 70%, Cubitainer® Package, 20 L
Documents & Certificates
Reviews
Q&A