Reagents EDTA Disodium Dihydrate, Purified, Poly Pail, 3 kg

Representative image only.

Reagents EDTA Disodium Dihydrate, Purified, Poly Pail, 3 kg

Reagents | Mfr: C2201100-3KG| Item: 6228501

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White solid
  • Density: 0.86
  • Boiling point: >252 °C (>485.6 °F), decomposes
  • Solubility information: 110 g/L at 20 °C (68 °F)
  • 3 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaC10H14N2O8Na2·2H2O
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • SynonymsEndrate, Edetate disodium, Disodium EDTA dihydrate, (Ethylenedinitrilo)tetraacetic acid, Disodium salt dihydrate
  • Weight (g)3000
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6381-92-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)252
  • Melting Point F (° F)485.6
  • DescriptionEDTA Disodium Dihydrate, Purified, Poly Pail, 3 kg
Documents & Certificates
Reviews
Q&A