Reagents Dimethyl Sulfoxide, ACS Reagent, Amber Glass Bottle, 4 L

Representative image only.

Reagents Dimethyl Sulfoxide, ACS Reagent, Amber Glass Bottle, 4 L

Reagents | Mfr: C1196650-4C| Item: 6228173

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.9%
  • Colorless liquid
  • Density: 1.1
  • Refractive index: 1.477 to 1.48nD
  • Solubility information: miscible
  • 4 L amber glasss bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(CH3)2SO
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDMSO
  • Volume (mL)4000
  • Viscosity (cSt)2.25
  • Formula Weight78.13
  • Vapor Pressure (torr)0.4153
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number67-68-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)189
  • Boiling Point F (° F)372.2
  • Melting Point C (° C)18.5
  • Melting Point F (° F)65.3
  • DescriptionDimethyl Sulfoxide, ACS Reagent, Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A