Reagents Deionized Water; MDA, APHA, EPA, AOAC Grade, Drum, 200 L

Representative image only.

Reagents Deionized Water; MDA, APHA, EPA, AOAC Grade, Drum, 200 L

Reagents | Mfr: C2346400-200E| Item: 6228624

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Water meets MDA, APHA, EPA, AOAC grade standards
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeMDA, APHA, EPA, AOAC
  • Empirical FormulaH2O
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDihydrogen oxide, Dihydrogen monoxide
  • Volume (mL)200000
  • Formula Weight18.02
  • Vapor Pressure (torr)23.8
  • PackagingDrum
  • DOT InformationNot regulated by DOT
  • Storage SymbolAmbient
  • CAS Number7732-18-5
  • Shelf Life (days)540
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionDeionized Water; MDA, APHA, EPA, AOAC Grade, Drum, 200 L
Documents & Certificates
Reviews
Q&A