Reagents Cyclohexane, ACS Reagent, Metal Can, 20 L

Representative image only.

Reagents Cyclohexane, ACS Reagent, Metal Can, 20 L

Reagents | Mfr: C1193500-20E6| Item: 6228169

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless liquid
  • Density: 0.78
  • Solubility information: 0.05 g/L at 20 °C (68 °F)
  • 20 L metal can

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6H12
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsHexahydrobenzene, Hexamethylene
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight84.16
  • Vapor Pressure (torr)77.256
  • PackagingMetal can
  • DOT InformationCYCLOHEXANE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number110-82-7
  • UN NumberUN1145
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)82.98
  • Boiling Point F (° F)181.36
  • Melting Point C (° C)-103.5
  • Melting Point F (° F)-154.3
  • Refractive Index1.426
  • DescriptionCyclohexane, ACS Reagent, Metal Can, 20 L
Documents & Certificates
Reviews
Q&A