Reagents Cobalt Chloride, Hexahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Cobalt Chloride, Hexahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1181000-100C2| Item: 6228165

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Pink to red solid
  • Density: 2.48
  • Solubility information: 767 g/L
  • pH range: 4.9
  • 100 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCoCl2·6H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsCobalt chloride 6-hydrate
  • Weight (g)100
  • Formula Weight237.93
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7791-13-1
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)1049
  • Boiling Point F (° F)1920.2
  • Melting Point C (° C)86
  • Melting Point F (° F)186.8
  • DescriptionCobalt Chloride, Hexahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
Documents & Certificates
Reviews
Q&A