Reagents Chloroform, ACS Reagent, Safety Coated Clear Glass Bottle, 2.5 L

Representative image only.

Reagents Chloroform, ACS Reagent, Safety Coated Clear Glass Bottle, 2.5 L

Reagents | Mfr: C1175500-2.5D| Item: 6228163

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.8%
  • Colorless liquid
  • Density: 1.48
  • Boiling point: 60.5 to 61.2 °C (140.9 to 142.16 °F)
  • Solubility information: 8 g/L at 20 °C (68 °F)
  • 2.5 L safety coated clear glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • ClassSolvents.
  • ChemistrySolvents
  • Volume (mL)2500
  • Formula Weight119.38
  • Vapor Pressure (torr)158.263
  • PackagingSafety coated clear glass bottle
  • DOT InformationCHLOROFORM, 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1888
  • CAS Number67-66-3
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-63.5
  • Melting Point F (° F)-82.3
  • Refractive Index1.446
  • DescriptionChloroform, ACS Reagent, Safety Coated Clear Glass Bottle, 2.5 L
Documents & Certificates
Reviews
Q&A