Reagents Ceric Ammonium Nitrate, ACS Reagent, Natural Poly Container, 2 kg

Representative image only.

Reagents Ceric Ammonium Nitrate, ACS Reagent, Natural Poly Container, 2 kg

Reagents | Mfr: C1171520-2A5| Item: 6228161

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98.5%
  • Orange-red or orange-yellow solid
  • Density: 1.1
  • Melting point: 107 to 108 °C (224.6 to 226.4 °F)
  • Solubility information: 1410 g/L at 20 °C (68 °F)
  • 2 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(NH4)2Ce(NO3)6
  • ClassSalts
  • ChemistryInorganics
  • SynonymsAmmonium cerium (IV) nitrate
  • FormSolid
  • Weight (g)2000
  • Formula Weight548.22
  • PackagingPoly container
  • DOT InformationNITRATES, INORGANIC, N.O.S. (CERIC AMMONIUM NITRATE), 5.1, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1477
  • CAS Number16774-21-3
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionCeric Ammonium Nitrate, ACS Reagent, Natural Poly Container, 2 kg
Documents & Certificates
Reviews
Q&A