Reagents CDTA, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents CDTA, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C1194010-25C1| Item: 6228170

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.5
  • Melting point: 213 to 216 °C (415.4 to 420.8 °F)
  • Solubility information: slightly soluble
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC14H22N2O8·H2O
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • Synonymstrans(1,2-Cyclohexylenedinitrilo)tetraacetic acid
  • Weight (g)25
  • Formula Weight364.35
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number13291-61-7
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionCDTA, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g
Documents & Certificates
Reviews
Q&A