Reagents Calcium Hydroxide, Purified, Fiber Drum, 50 lb

Representative image only.

Reagents Calcium Hydroxide, Purified, Fiber Drum, 50 lb

Reagents | Mfr: C2164500-50F5| Item: 6228477

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 95%
  • Colorless/white solid
  • Density: 2.24
  • Boiling point: 2850 °C (5162 °F) with decomposition
  • Melting point: 580 °C (1076 °F) with decomposition
  • Solubility information: 1.6 g/L at 20 °C (68 °F)
  • 50 lb fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaCa(OH)2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsSlaked lime, Calcium hydrate
  • Weight (g)22680
  • Formula Weight74.09
  • Vapor Pressure (torr)< 0.7501
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1305-62-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionCalcium Hydroxide, Purified, Fiber Drum, 50 lb
Documents & Certificates
Reviews
Q&A