Reagents Calcium Chloride Dihydrate, Technical, Poly Pail, 10 kg

Representative image only.

Reagents Calcium Chloride Dihydrate, Technical, Poly Pail, 10 kg

Reagents | Mfr: C2163700-10KG| Item: 6228476

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.84
  • Boiling point: >1600 °C (>2912 °F)
  • Solubility information: 977 g/L at 0 °C (32 °F)
  • pH range: 4.5 to 8
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaCaCl2·2H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsCalcium chloride 2-hydrate
  • Weight (g)10000
  • Formula Weight147.01
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10035-04-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)176
  • Melting Point F (° F)348.8
  • DescriptionCalcium Chloride Dihydrate, Technical, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A