Reagents Calcium Chloride Anhydrous, Poly Pail, 10 kg

Representative image only.

Reagents Calcium Chloride Anhydrous, Poly Pail, 10 kg

Reagents | Mfr: C2162550-10KG| Item: 6228474

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.15
  • Mesh size: 4 to 8
  • Solubility information: 740 g/L at 20 °C (68 °F)
  • pH range: 8 to 10
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaCaCl2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight110.98
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10043-52-4
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)1670
  • Boiling Point F (° F)3038
  • Melting Point C (° C)772
  • Melting Point F (° F)1421.6
  • DescriptionCalcium Chloride Anhydrous, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A