Reagents Bromothymol Blue Indicator, 0.04% (w/v), Volumetric, Natural Poly Bottle, 100 mL

Representative image only.

Reagents Bromothymol Blue Indicator, 0.04% (w/v), Volumetric, Natural Poly Bottle, 100 mL

Reagents | Mfr: C7107000-100A| Item: 6228767

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Aqueous
  • Dark colored liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 6.0 (yellow) to 7.6 (blue)
  • 100 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.04% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)100
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionBromothymol Blue Indicator, 0.04% (w/v), Volumetric, Natural Poly Bottle, 100 mL
Documents & Certificates
Reviews
Q&A