Reagents Bromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Bromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Natural Poly Bottle, 500 mL

Reagents | Mfr: C7106300-500A| Item: 6228766

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Aqueous
  • Dark reddish-purple liquid
  • Density: 1
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 3.0 (yellow) to 4.6 (blue)
  • 500 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.04% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number62625-28-9, 7732-18-5
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionBromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Natural Poly Bottle, 500 mL
Documents & Certificates
Reviews
Q&A