Reagents Bromocresol Purple Indicator, 0.04% (w/v), Volumetric, Cubitainer® Package, 10 L

Representative image only.

Reagents Bromocresol Purple Indicator, 0.04% (w/v), Volumetric, Cubitainer® Package, 10 L

Reagents | Mfr: C7106000-10F| Item: 6228764

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Purple liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 5.2 (yellow) to 6.8 (purple)
  • 10 L Cubitainer® package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.04% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer® package
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number115-40-2, 1310-73-2, 7732-18-5
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionBromocresol Purple Indicator, 0.04% (w/v), Volumetric, Cubitainer® Package, 10 L
Documents & Certificates
Reviews
Q&A