Reagents Bromocresol Purple, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g

Representative image only.

Reagents Bromocresol Purple, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g

Reagents | Mfr: C6106000-5C2| Item: 6228659

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Light yellow-tan solid
  • Density: 0.991
  • Melting point: 240 °C (464 °C), decomposes
  • Solubility information: slightly soluble
  • pH range: 5.2 (yellow) to 6.8 (purple)
  • 5 g amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)5
  • PackagingWide-Mouth Amber Glass Bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number115-40-2
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionBromocresol Purple, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g
Documents & Certificates
Reviews
Q&A