Reagents Boric Acid, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Boric Acid, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C1104300-10KG| Item: 6228114

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white crystal
  • Melting point: 168 to 170 °C (334.4 to 338 °F)
  • Density: 1.44
  • Solubility information: 47.5 g/L at 20 °C (68 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaH3BO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight61.83
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10043-35-3
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)300
  • Boiling Point F (° F)572
  • Refractive Index1.44
  • DescriptionBoric Acid, ACS Reagent, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A