Reagents Barium Chloride Dihydrate, Poly Pail, 10 kg

Representative image only.

Reagents Barium Chloride Dihydrate, Poly Pail, 10 kg

Reagents | Mfr: C2147000-10KG| Item: 6228393

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 3.86
  • Solubility information: 370 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 8.2
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaBaCl2·2H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsBarium chloride 2-hydrate
  • FormSolid
  • Weight (g)10000
  • Formula Weight244.26
  • PackagingPoly pail
  • DOT InformationBARIUM COMPOUNDS, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1564
  • CAS Number10326-27-9
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)1560
  • Boiling Point F (° F)2840
  • Melting Point C (° C)963
  • Melting Point F (° F)1765.4
  • DescriptionBarium Chloride Dihydrate, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A