Reagents Barium Chloride, 25% (w/v) as BaCl2, Standardized, Cubitainer® Package, 10 L

Representative image only.

Reagents Barium Chloride, 25% (w/v) as BaCl2, Standardized, Cubitainer® Package, 10 L

Reagents | Mfr: CB111200-10F| Item: 6229005

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Aqueous
  • Colorless liquid
  • Density: 1.2
  • Specific gravity: 1.1996 to 1.2278 @ 20 °C (68 °F)
  • Solubility information: miscible
  • pH range: 5.2 to 8.2
  • 10 L Cubitainer package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration 25% w/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer
  • DOT InformationTOXIC LIQUID, INORGANIC, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3287
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Boiling Point C (° C)101
  • Boiling Point F (° F)213.8
  • DescriptionBarium Chloride, 25% (w/v) as BaCl2, Standardized, Cubitainer® Package, 10 L
Documents & Certificates
Reviews
Q&A