Reagents Barium Chloride, 20% (w/v), Volumetric, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Barium Chloride, 20% (w/v), Volumetric, Natural Poly Bottle, 500 mL

Reagents | Mfr: CB100500-500A| Item: 6228999

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Aqueous
  • Colorless liquid
  • Density: 1.16
  • Boiling point: approximately 101 °C (213.8 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 5.2 to 8.2
  • 500 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration20% w/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationTOXIC LIQUID, INORGANIC, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3287
  • Shelf Life (days)1095
  • Shelf Life (months)36
  • DescriptionBarium Chloride, 20% (w/v), Volumetric, Natural Poly Bottle, 500 mL
Documents & Certificates
Reviews
Q&A