Reagents Ammonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Ammonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 500 mL

Reagents | Mfr: CA010300-500A| Item: 6228889

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • N/10
  • Assay 0.0995 to 0.1005 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 4.5 to 6
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Normality0.1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionAmmonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A