Reagents Ammonium Sulfate, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Ammonium Sulfate, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C1138000-10KG| Item: 6228151

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white solid
  • Density: 1.77
  • Solubility information: 754 g/L at 20 °C (68 °F)
  • pH range: 5 to 6
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(NH4)2SO4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsAmmonium tetraoxosulfate (VI)
  • FormSolid
  • Weight (g)10000
  • Formula Weight132.14
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7783-20-2
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)280
  • Melting Point F (° F)536
  • DescriptionAmmonium Sulfate, ACS Reagent, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A