Reagents Ammonium Persulfate, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Ammonium Persulfate, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1136500-12KG| Item: 6228148

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white solid
  • Density: 1.98
  • Melting point: 119 to 160 °C (264.2 to 320 °F), decomposes
  • Solubility information: 559 g/L at 20 °C (68 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(NH4)2S2O8
  • ClassSalts
  • ChemistryInorganics
  • SynonymsAmmonium peroxodisulfate, Peroxidisulfuric acid diammonium salt
  • FormSolid
  • Weight (g)12000
  • Formula Weight228.2
  • PackagingPoly pail
  • DOT InformationAMMONIUM PERSULFATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number7727-54-0
  • UN NumberUN1444
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionAmmonium Persulfate, ACS Reagent, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A