Reagents Ammonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Ammonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C2136000-100C2| Item: 6228391

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.5
  • Solubility information: 45 g/L at 20 °C (68 °F)
  • 100 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear Formula(COONH4)2·H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsAmmonium oxalate monohydrate
  • Weight (g)100
  • Formula Weight142.11
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6009-70-7
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Melting Point C (° C)70
  • Melting Point F (° F)158
  • DescriptionAmmonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 100 g
Documents & Certificates
Reviews
Q&A