Reagents Ammonium Nitrate, ACS Reagent, Poly Pail, 3 kg

Representative image only.

Reagents Ammonium Nitrate, ACS Reagent, Poly Pail, 3 kg

Reagents | Mfr: C1135500-3KG| Item: 6228145

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white solid
  • Density: 1.72
  • Flash point: >93.3
  • Solubility information: 1920 g/L at 20 °C (68 °F)
  • pH range: 4.5 to 6
  • 3 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNH4NO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • Formula Weight80.04
  • PackagingPoly pail
  • DOT InformationAMMONIUM NITRATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number6484-52-2
  • UN NumberUN1942
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)210
  • Boiling Point F (° F)410
  • Melting Point C (° C)170
  • Melting Point F (° F)338
  • DescriptionAmmonium Nitrate, ACS Reagent, Poly Pail, 3 kg
Documents & Certificates
Reviews
Q&A