Reagents Ammonium Molybdate, ACS Reagent, Wide-Mouth Natural Poly Container, 3 kg

Representative image only.

Reagents Ammonium Molybdate, ACS Reagent, Wide-Mouth Natural Poly Container, 3 kg

Reagents | Mfr: C1135000-3A5| Item: 6228143

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 81%
  • White to slightly yellow-green solid
  • Density: 2.5
  • Solubility information: 430 g/L
  • 3 kg wide-mouth natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(NH4)6Mo7O24·4H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsAmmonium molybdate tetrahydrate, Ammonium docosaoxoheptamolybdate(6–)
  • FormSolid
  • Weight (g)3000
  • Formula Weight1236
  • PackagingWide-mouth natural poly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number12054-85-2
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)190
  • Boiling Point F (° F)374
  • Melting Point C (° C)190
  • Melting Point F (° F)374
  • DescriptionAmmonium Molybdate, ACS Reagent, Wide-Mouth Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A