Reagents Ammonium Hydroxide, 25% (v/v), Volumetric, Natural Poly Bottle, 1 L

Representative image only.

Reagents Ammonium Hydroxide, 25% (v/v), Volumetric, Natural Poly Bottle, 1 L

Reagents | Mfr: CA100300-1A| Item: 6228947

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 1:3
  • Aqueous
  • Colorless liquid
  • Density: 0.98
  • Boiling point: approximately 36 °C (96.8 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 11.9
  • 1 L natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration25% v/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly bottle
  • DOT InformationCORROSIVE LIQUID, BASIC, INORGANIC, N.O.S. (AMMONIUM HYDROXIDE), 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3266
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionAmmonium Hydroxide, 25% (v/v), Volumetric, Natural Poly Bottle, 1 L
Documents & Certificates
Reviews
Q&A